siRNA Details
Gene Symbol: BRPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319049.1 | NP_001305978.1 | 6, 7 | 2411 | ||
NM_001319050.1 | NP_001305979.1 | 6, 7 | 2411 | |||
NM_004634.2 | NP_004625.2 | 6, 7 | 2390 | |||
XM_005265449.1 | XP_005265506.1 | 6, 7 | 2411 | |||
XM_011534101.1 | XP_011532403.1 | 6, 7 | 2411 | |||
XM_011534102.1 | XP_011532404.1 | 6, 7 | 2209 | |||
GenBank mRNA | AK293865.1 | - | 6, 7 | 2411 | ||
AL713696.1 | - | 6, 7 | 2220 | |||
M91585.1 | - | 6, 7 | 2019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
