siRNA Details
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035247.2 | NP_001030324.2 | 3, 4 | 308 | ||
NM_005659.6 | NP_005650.2 | 3, 4 | 308 | |||
XM_011530361.2 | XP_011528663.1 | 3, 4 | 434 | |||
GenBank mRNA | AF141201.1 | - | 2, 3 | 178 | ||
AJ239058.1 | - | 2, 3 | 178 | |||
AK225877.1 | - | 3, 4 | 265 | |||
AK304734.1 | - | 3, 4 | 278 | |||
AK308786.1 | - | 3, 4 | 252 | |||
AK310950.1 | - | 3, 4 | 260 | |||
AY101594.1 | - | 3, 4 | 235 | |||
BC001049.2 | - | 3, 4 | 255 | 3507963 | ||
BC005087.1 | - | 3, 4 | 256 | 3830774 | ||
BX648483.1 | - | 3, 4 | 240 | |||
CB128364.1 | - | 3, 4 | 308 | |||
CR456607.1 | - | 3, 4 | 192 | |||
DB496066.1 | - | 3, 4 | 292 | |||
U64444.1 | - | 3, 4 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
