siRNA Details
Gene Symbol: TBCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005993.4 | NP_005984.3 | 1, 2 | 312 | ||
XM_006722290.2 | XP_006722353.1 | 1, 2 | 290 | |||
XM_011523590.1 | XP_011521892.1 | 1, 2 | 290 | |||
XM_017024988.1 | XP_016880477.1 | 1, 2 | 290 | |||
GenBank mRNA | AB023205.1 | - | 1, 2 | 291 | ||
AJ006417.1 | - | 1, 2 | 224 | |||
AK293471.1 | - | 1, 2 | 290 | |||
BC003094.1 | - | 1, 2 | 312 | 3503192 | ||
BC006364.1 | - | 1, 2 | 269 | 4131114 | ||
BI464569.1 | - | 1, 2 | 308 | |||
CA390153.1 | - | 2, 3 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
