siRNA Details
Gene Symbol: TCHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007113.3 | NP_009044.2 | 2 | 179 | ||
GenBank mRNA | AK307946.1 | - | 355 | |||
AY827491.1 | - | 2 | 169 | |||
BC131826.1 | - | 1 | 1543 | |||
BC131827.1 | - | 1 | 1543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
