siRNA Details
Gene Symbol: SLMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF100750.1 | - | 2, 3 | 210 | ||
AK302934.1 | - | 2, 3 | 141 | |||
AK304493.1 | - | 2, 3 | 193 | |||
BC115701.1 | - | 14, 15 | 1482 | 40081893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
