siRNA Details
Gene Symbol: ZNF225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321685.1 | NP_001308614.1 | 6 | 1336 | ||
NM_013362.3 | NP_037494.2 | 5 | 1171 | |||
XM_005259223.3 | XP_005259280.1 | 2 | 717 | |||
XM_011527285.2 | XP_011525587.1 | 5 | 1191 | |||
XM_011527286.2 | XP_011525588.1 | 5 | 1835 | |||
GenBank mRNA | AF187991.1 | - | 5 | 1145 | ||
AK223477.1 | - | 5 | 1090 | |||
AK292437.1 | - | 5 | 1175 | |||
AK313301.1 | - | 4 | 1178 | |||
BC108912.2 | - | 4 | 968 | 40027157 |
Gene Symbol: ZNF155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA164828.1 | - | 1 | 480 |
