siRNA Details
Gene Symbol: HIST1H2AJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021066.2 | NP_066544.1 | 1 | 380 | ||
GenBank mRNA | BC066232.1 | - | 1 | 380 | ||
BC066233.1 | - | 1 | 380 | |||
BC066234.1 | - | 1 | 380 | |||
BC066235.1 | - | 1 | 380 | |||
BC066236.1 | - | 1 | 380 | |||
BC066237.1 | - | 1 | 380 | |||
BC133048.1 | - | 1 | 469 | 40147070 | ||
BC133050.1 | - | 1 | 469 | 40147072 |
