siRNA Details
Gene Symbol: TCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030752.2 | NP_110379.2 | 2 | 319 | ||
GenBank mRNA | AK312604.1 | - | 2 | 288 | ||
BC000665.1 | - | 2 | 209 | 3349237 | ||
BM827784.1 | - | 2 | 325 | |||
BT006969.1 | - | 2 | 84 | |||
X52882.1 | - | 2 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
