siRNA Details
Gene Symbol: SH2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291424.1 | NP_001278353.1 | 7 | 1766 | ||
NM_005475.2 | NP_005466.1 | 8 | 2689 | |||
XM_005253818.4 | XP_005253875.1 | 9 | 2833 | |||
XM_005253819.4 | XP_005253876.1 | 8 | 2716 | |||
XM_006719180.3 | XP_006719243.1 | 7 | 2318 | |||
XM_011537719.2 | XP_011536021.1 | 9 | 2836 | |||
XM_011537720.2 | XP_011536022.1 | 9 | 2501 | |||
XM_011537721.2 | XP_011536023.1 | 7 | 4301 | |||
GenBank mRNA | AB208911.1 | - | 7 | 1925 | ||
AF055581.1 | - | 8 | 2689 | |||
AK025745.1 | - | 1 | 535 | |||
AK302173.1 | - | 7 | 1765 | |||
BC136451.1 | - | 8 | 2375 | 9020440 |
