siRNA Details
Gene Symbol: PRELID3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142405.1 | NP_001135877.1 | 7 | 1611 | ||
NM_001142406.1 | NP_001135878.1 | 6 | 1635 | |||
NM_006553.3 | NP_006544.2 | 7 | 1608 | |||
GenBank mRNA | AF048849.1 | - | 1 | 541 | ||
AK056046.1 | - | 7 | 1562 | |||
AL359942.1 | - | 5 | 1593 | |||
BC007528.1 | - | 2 | 1017 | |||
CR627465.1 | - | 5 | 1593 |
