siRNA Details
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 10, 11 | 1711 | ||
GenBank mRNA | AB621816.1 | - | 10, 11 | 1778 | ||
AF143946.1 | - | 6, 7 | 945 | |||
AK023808.1 | - | 3, 4 | 425 | |||
AK128030.1 | - | 10, 11 | 1711 | |||
AU140541.2 | - | 3, 4 | 425 | |||
BC025284.1 | - | 7, 8 | 1188 | |||
BC063578.1 | - | 8, 9 | 1505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
