siRNA Details
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 16 | 2846 | ||
GenBank mRNA | AB621816.1 | - | 16 | 2913 | ||
AF143946.1 | - | 12 | 2080 | |||
AK023808.1 | - | 9 | 1559 | |||
AK128030.1 | - | 16 | 2846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
