siRNA Details
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 27 | 6459 | ||
GenBank mRNA | AF143946.1 | - | 22 | 5507 | ||
AK128030.1 | - | 27 | 6459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
