siRNA Details
Gene Symbol: PPIP5K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130858.2 | NP_001124330.1 | 28 | 3423 | ||
XM_005254803.1 | XP_005254860.1 | 28 | 3411 | |||
XM_011522249.1 | XP_011520551.1 | 29 | 3561 | |||
XM_011522250.1 | XP_011520552.1 | 29 | 3664 | |||
XM_011522252.1 | XP_011520554.1 | 28 | 3535 | |||
XM_011522254.1 | XP_011520556.1 | 28 | 3431 | |||
XM_011522258.1 | XP_011520560.1 | 23 | 2821 | |||
XM_011522259.1 | XP_011520561.1 | 28 | 3387 | |||
XM_017022735.1 | XP_016878224.1 | 28 | 3878 | |||
XM_017022736.1 | XP_016878225.1 | 28 | 3846 | |||
XM_017022737.1 | XP_016878226.1 | 28 | 3527 | |||
XM_017022738.1 | XP_016878227.1 | 28 | 3451 | |||
XM_017022740.1 | XP_016878229.1 | 28 | 3514 | |||
XM_017022741.1 | XP_016878230.1 | 27 | 3727 | |||
XM_017022742.1 | XP_016878231.1 | 28 | 3519 | |||
XM_017022743.1 | XP_016878232.1 | 27 | 3385 | |||
XM_017022744.1 | XP_016878233.1 | 27 | 3326 | |||
XM_017022745.1 | XP_016878234.1 | 28 | 3490 | |||
XM_017022746.1 | XP_016878235.1 | 27 | 3361 | |||
XM_017022760.1 | XP_016878249.1 | 27 | 4859 | |||
GenBank mRNA | AF502586.1 | - | 27 | 3301 | ||
AF502589.1 | - | 26 | 3030 | |||
AF543190.1 | - | 26 | 2980 | |||
BC057395.1 | - | 27 | 3369 | 6142663 | ||
BX647870.1 | - | 5 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
