siRNA Details
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 10 | 906 | ||
NR_027410.1 | 18 | 3154 | ||||
XM_011521596.1 | XP_011519898.1 | 17 | 1751 | |||
XM_011521597.1 | XP_011519899.1 | 16 | 1702 | |||
XM_017022210.1 | XP_016877699.1 | 18 | 2055 | |||
XM_017022211.1 | XP_016877700.1 | 18 | 1824 | |||
XM_017022212.1 | XP_016877701.1 | 17 | 2378 | |||
XM_017022213.1 | XP_016877702.1 | 17 | 1780 | |||
XM_017022214.1 | XP_016877703.1 | 18 | 2055 | |||
XM_017022215.1 | XP_016877704.1 | 18 | 2577 | |||
XM_017022216.1 | XP_016877705.1 | 17 | 3175 | |||
XM_017022217.1 | XP_016877706.1 | 16 | 3097 | |||
XM_017022218.1 | XP_016877707.1 | 18 | 3204 | |||
GenBank mRNA | AK097551.1 | - | 10 | 895 | ||
BC143254.1 | - | 10 | 852 | 9051756 | ||
BC144610.1 | - | 8 | 643 | 9053141 | ||
BX647673.1 | - | 17 | 3123 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 10 | 906 | ||
NR_027409.1 | 18 | 3168 | ||||
XM_006720440.2 | XP_006720503.1 | 10 | 874 | |||
XM_017022018.1 | XP_016877507.1 | 14 | 3302 | |||
XM_017022019.1 | XP_016877508.1 | 13 | 2998 | |||
XM_017022020.1 | XP_016877509.1 | 8 | 996 | |||
GenBank mRNA | AB020662.1 | - | 9 | 886 | ||
AF163441.1 | - | 8 | 1330 | |||
AF204231.1 | - | 9 | 1223 | |||
AL137686.1 | - | 8 | 630 | |||
BC093673.1 | - | 10 | 852 | 7939518 | ||
BC104800.1 | - | 10 | 852 | 8143803 | ||
BC144609.1 | - | 8 | 643 | 9053140 | ||
BC150329.1 | - | 9 | 897 | |||
BC152410.1 | - | 9 | 886 | |||
BX648160.1 | - | 17 | 3122 | |||
BX648575.1 | - | 17 | 2875 | |||
CR749361.1 | - | 10 | 2059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
