siRNA Details
Gene Symbol: CEP83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042399.1 | NP_001035858.1 | 16 | 2900 | ||
NM_016122.2 | NP_057206.2 | 17 | 2953 | |||
XM_011538424.1 | XP_011536726.1 | 18 | 2999 | |||
XM_011538427.2 | XP_011536729.1 | 17 | 2725 | |||
XM_011538429.2 | XP_011536731.1 | 13 | 2196 | |||
XM_017019387.1 | XP_016874876.1 | 16 | 2704 | |||
XM_017019388.1 | XP_016874877.1 | 16 | 4569 | |||
XM_017019389.1 | XP_016874878.1 | 18 | 2779 | |||
GenBank mRNA | AF155115.1 | - | 17 | 2667 |
