siRNA Details
Gene Symbol: NT5DC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031701.2 | NP_001026871.1 | 14 | 3829 | ||
GenBank mRNA | AB032773.1 | - | 14 | 3632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
