siRNA Details
Gene Symbol: LRRC37A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002940.2 | 1 | 1133 | |||
GenBank mRNA | AK000982.1 | - | 3 | 1025 | ||
JQ790585.1 | - | 6 | 594 | |||
JQ790586.1 | - | 6 | 594 | |||
JQ790587.1 | - | 4 | 488 | |||
JQ790588.1 | - | 4 | 488 | |||
JQ790589.1 | - | 4 | 488 | |||
JQ790600.1 | - | 5 | 560 | |||
JQ790630.1 | - | 6 | 594 | |||
JQ790631.1 | - | 6 | 632 | |||
JQ790643.1 | - | 4 | 488 | |||
JQ790644.1 | - | 4 | 488 | |||
JQ790645.1 | - | 4 | 488 | |||
JQ790646.1 | - | 4 | 488 | |||
JQ790647.1 | - | 4 | 488 | |||
JQ790648.1 | - | 4 | 488 | |||
JQ790649.1 | - | 4 | 488 | |||
JQ790650.1 | - | 4 | 488 | |||
JQ790651.1 | - | 4 | 488 | |||
JQ790652.1 | - | 4 | 488 | |||
JQ790653.1 | - | 4 | 487 | |||
JQ790654.1 | - | 4 | 488 | |||
JQ790655.1 | - | 4 | 488 | |||
JQ790656.1 | - | 4 | 488 | |||
JQ790657.1 | - | 4 | 488 | |||
JQ790658.1 | - | 4 | 488 | |||
JQ790659.1 | - | 4 | 488 | |||
JQ790666.1 | - | 3 | 369 | |||
JQ790681.1 | - | 5 | 560 | |||
JQ790682.1 | - | 5 | 560 | |||
JQ790683.1 | - | 5 | 560 | |||
JQ790684.1 | - | 5 | 560 | |||
JQ790685.1 | - | 5 | 560 | |||
JQ790686.1 | - | 5 | 560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
