siRNA Details
Gene Symbol: AMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257360.1 | NP_001244289.1 | 2 | 772 | ||
NM_001308170.1 | NP_001295099.1 | 1 | 306 | |||
NM_004037.7 | NP_004028.3 | 1 | 639 | |||
GenBank mRNA | AK302939.1 | - | 1 | 148 | ||
AK314494.1 | - | 2 | 695 | |||
U16268.1 | - | 1 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
