siRNA Details
Gene Symbol: MCTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159643.1 | NP_001153115.1 | 9, 10 | 1374 | ||
NM_001159644.1 | NP_001153116.1 | 2, 3 | 441 | |||
NM_018349.3 | NP_060819.3 | 9, 10 | 1374 | |||
XM_005254955.3 | XP_005255012.1 | 10, 11 | 1855 | |||
XM_005254960.2 | XP_005255017.1 | 2, 3 | 446 | |||
XM_006720603.2 | XP_006720666.1 | 11, 12 | 1611 | |||
XM_011521770.1 | XP_011520072.1 | 9, 10 | 1374 | |||
XM_011521771.2 | XP_011520073.1 | 11, 12 | 1780 | |||
XM_011521772.2 | XP_011520074.1 | 10, 11 | 1670 | |||
XM_011521773.2 | XP_011520075.1 | 9, 10 | 1374 | |||
XM_011521774.2 | XP_011520076.1 | 9, 10 | 1374 | |||
XM_011521775.2 | XP_011520077.1 | 3, 4 | 1126 | |||
XM_017022403.1 | XP_016877892.1 | 8, 9 | 1338 | |||
XM_017022404.1 | XP_016877893.1 | 11, 12 | 1780 | |||
XM_017022405.1 | XP_016877894.1 | 10, 11 | 1785 | |||
GenBank mRNA | AK002037.1 | - | 2, 3 | 149 | ||
AK090622.1 | - | 10, 11 | 1463 | |||
BC025708.1 | - | 2, 3 | 402 | |||
BC111024.1 | - | 2, 3 | 274 | |||
BC131527.1 | - | 9, 10 | 1309 | 40035019 | ||
DA862691.1 | - | 2, 3 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
