siRNA Details
Gene Symbol: MCTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159643.1 | NP_001153115.1 | 20 | 3428 | ||
NM_018349.3 | NP_060819.3 | 22 | 3593 | |||
XM_005254955.3 | XP_005255012.1 | 23 | 4074 | |||
XM_005254960.2 | XP_005255017.1 | 15 | 2665 | |||
XM_006720603.2 | XP_006720666.1 | 24 | 3830 | |||
XM_011521770.1 | XP_011520072.1 | 21 | 3470 | |||
XM_011521771.2 | XP_011520073.1 | 24 | 3999 | |||
XM_011521772.2 | XP_011520074.1 | 23 | 3889 | |||
XM_011521775.2 | XP_011520077.1 | 16 | 3345 | |||
XM_017022403.1 | XP_016877892.1 | 21 | 3557 | |||
XM_017022404.1 | XP_016877893.1 | 22 | 3834 | |||
XM_017022405.1 | XP_016877894.1 | 21 | 3839 | |||
GenBank mRNA | AK001953.1 | - | 6 | 1412 | ||
AK002037.1 | - | 15 | 2368 | |||
AL360258.1 | - | 1 | 1672 | |||
AL697863.1 | - | 1 | 161 | |||
BC041387.1 | - | 24 | 4015 | 5276211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
