siRNA Details
Gene Symbol: PRR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018457.3 | NP_060927.1 | 3 | 263 | ||
GenBank mRNA | AF217517.1 | - | 1 | 176 | ||
AK074398.1 | - | 1 | 159 | |||
BC014257.2 | - | 1 | 163 | 4765821 | ||
BC016064.1 | - | 1 | 160 | 4852862 | ||
BC021019.1 | - | 1 | 132 | |||
BC066943.1 | - | 1 | 162 | 4817871 | ||
BC071979.1 | - | 1 | 106 | |||
CN392914.1 | - | 3 | 277 | |||
CR533459.1 | - | 2 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
