siRNA Details
Gene Symbol: ADPRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020233.4 | NP_064618.3 | 3 | 737 | ||
XM_005256738.2 | XP_005256795.1 | 3 | 737 | |||
GenBank mRNA | AB073393.1 | - | 3 | 708 | ||
BC001294.2 | - | 2 | 780 | 3452502 | ||
BC070155.1 | - | 3 | 820 | 4656998 | ||
KF880964.1 | - | 2 | 662 |
