siRNA Details
Gene Symbol: MAPK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315.2 | NP_001306.1 | 12 | 2737 | ||
NM_139012.2 | NP_620581.1 | 12 | 2737 | |||
NM_139014.2 | NP_620583.1 | 11 | 2658 | |||
XM_006714998.3 | XP_006715061.1 | 12 | 2212 | |||
XM_011514310.2 | XP_011512612.2 | 12 | 3005 | |||
GenBank mRNA | BC031574.1 | - | 12 | 2593 | 5181064 | |
L35263.1 | - | 12 | 2618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
