siRNA Details
Gene Symbol: C17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022344.3 | NP_071739.2 | 1, 2 | 215 | ||
XM_005258022.3 | XP_005258079.1 | 2, 3 | 233 | |||
GenBank mRNA | AF052093.1 | - | 1, 2 | 142 | ||
AF305686.1 | - | 1, 2 | 171 | |||
BC026017.2 | - | 1, 2 | 161 | 3352761 | ||
BC035715.1 | - | 1, 2 | 169 | 5588164 | ||
BC047325.1 | - | 1, 2 | 155 | 5493477 | ||
BC054035.1 | - | 1, 2 | 149 | 6143106 | ||
BP308900.1 | - | 1, 2 | 214 | |||
CD706845.1 | - | 1, 2 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
