siRNA Details
Gene Symbol: FAM173A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271285.1 | NP_001258214.1 | 4 | 809 | ||
NM_023933.2 | NP_076422.1 | 5 | 860 | |||
GenBank mRNA | AK123186.1 | - | 3 | 1673 | ||
BC001181.1 | - | 5 | 844 | 3355322 |
