siRNA Details
Gene Symbol: GNPTAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024312.4 | NP_077288.2 | 13 | 2047 | ||
XM_006719593.2 | XP_006719656.1 | 13 | 2042 | |||
XM_011538731.2 | XP_011537033.1 | 13 | 2043 | |||
XM_017019961.1 | XP_016875450.1 | 13 | 1944 | |||
XM_017019962.1 | XP_016875451.1 | 13 | 2149 | |||
GenBank mRNA | AB033034.2 | - | 6 | 938 | ||
AK056137.1 | - | 13 | 1918 | |||
AM085438.1 | - | 13 | 2034 | |||
AY687932.1 | - | 13 | 2019 | |||
BC131787.1 | - | 13 | 1975 | 40126580 |
