siRNA Details
Gene Symbol: RNASEH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142279.2 | NP_001135751.1 | 6, 7 | 917 | ||
NM_024570.3 | NP_078846.2 | 6, 7 | 917 | |||
XM_006719867.3 | XP_006719930.1 | 6, 7 | 606 | |||
XM_011535229.1 | XP_011533531.1 | 6, 7 | 839 | |||
XM_011535230.2 | XP_011533532.1 | 6, 7 | 839 | |||
XM_011535231.2 | XP_011533533.1 | 6, 7 | 839 | |||
XM_011535233.2 | XP_011533535.1 | 7, 8 | 792 | |||
XM_017020747.1 | XP_016876236.1 | 6, 7 | 839 | |||
GenBank mRNA | AK021774.1 | - | 6, 7 | 804 | ||
AK124228.1 | - | 1, 2 | 2583 | |||
AK223340.1 | - | 6, 7 | 804 | |||
AY764036.1 | - | 6, 7 | 825 | |||
BC001397.2 | - | 6, 7 | 627 | |||
BC005088.1 | - | 6, 7 | 627 | |||
BC007332.2 | - | 6, 7 | 627 | |||
BC010174.2 | - | 6, 7 | 627 | |||
BC036744.1 | - | 6, 7 | 788 | 5195426 | ||
BX328027.1 | - | 3, 4 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
