siRNA Details
Gene Symbol: RHBDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005498.3 | NP_001005498.2 | 18 | 3279 | ||
NM_024599.5 | NP_078875.4 | 19 | 3441 | |||
XM_005257669.3 | XP_005257726.1 | 19 | 3802 | |||
XM_005257670.1 | XP_005257727.1 | 19 | 3311 | |||
XM_011525249.2 | XP_011523551.1 | 19 | 4758 | |||
XM_011525250.2 | XP_011523552.1 | 20 | 3468 | |||
XM_011525251.2 | XP_011523553.1 | 19 | 3285 | |||
XM_017025079.1 | XP_016880568.1 | 19 | 3715 | |||
GenBank mRNA | AK074034.1 | - | 16 | 4379 | ||
AK225684.1 | - | 14 | 2418 | |||
AL833680.1 | - | 12 | 2499 | |||
BC016034.1 | - | 19 | 3421 | 3910543 | ||
BC035829.1 | - | 14 | 2527 | |||
BM801831.1 | - | 1 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
