siRNA Details
Gene Symbol: TEFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024683.3 | NP_078959.3 | 4 | 902 | ||
XM_006722084.1 | XP_006722147.1 | 4 | 1386 | |||
GenBank mRNA | AF086465.1 | - | 1 | 140 | ||
AJ314645.1 | - | 4 | 869 | |||
AK026382.1 | - | 4 | 828 | |||
AK225474.1 | - | 4 | 828 | |||
BC009025.2 | - | 3 | 802 | |||
BC073169.1 | - | 3 | 525 | |||
CR457360.1 | - | 2 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
