siRNA Details
Gene Symbol: DHRS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024705.2 | NP_078981.1 | 8 | 1153 | ||
GenBank mRNA | AK023701.1 | - | 8 | 1148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
