siRNA Details
Gene Symbol: ACSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288968.1 | NP_001275897.1 | 12 | 1482 | ||
NM_001288969.1 | NP_001275898.1 | 11 | 1368 | |||
NM_001288970.1 | NP_001275899.1 | 10 | 1278 | |||
NM_001288971.1 | NP_001275900.1 | 9 | 1082 | |||
NM_001288972.1 | NP_001275901.1 | 11 | 1324 | |||
NM_025149.5 | NP_079425.3 | 11 | 1407 | |||
NR_110232.1 | 2 | 192 | ||||
XM_006722110.3 | XP_006722173.1 | 11 | 1347 | |||
XM_011525294.1 | XP_011523596.1 | 11 | 1329 | |||
GenBank mRNA | AK024573.1 | - | 11 | 1314 | ||
AK225044.1 | - | 11 | 1314 | |||
AK225561.1 | - | 11 | 1313 | |||
AK294205.1 | - | 12 | 1424 | |||
AK295258.1 | - | 11 | 1276 | |||
AK300145.1 | - | 10 | 1186 | |||
AK300625.1 | - | 9 | 1082 | |||
AK308584.1 | - | 2 | 178 | |||
AK311109.1 | - | 2 | 192 | |||
AY358660.1 | - | 11 | 1343 | |||
BC012053.1 | - | 11 | 1320 | 4577089 | ||
BC014123.2 | - | 11 | 1327 | 4560029 |
