siRNA Details
Gene Symbol: FAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031208.3 | NP_112485.1 | 1 | 734 | ||
GenBank mRNA | AK094199.1 | - | 1 | 733 | ||
AL136720.1 | - | 1 | 734 | |||
CR533499.1 | - | 1 | 728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
