siRNA Details
Gene Symbol: SNX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032167.4 | NP_115543.3 | 8 | 921 | ||
XM_005255682.3 | XP_005255739.1 | 8 | 946 | |||
XM_005255683.3 | XP_005255740.1 | 8 | 962 | |||
XM_011522738.2 | XP_011521040.1 | 8 | 1093 | |||
XM_011522741.2 | XP_011521043.1 | 8 | 1077 | |||
XM_011522743.2 | XP_011521045.1 | 8 | 1081 | |||
XM_011522744.2 | XP_011521046.1 | 8 | 952 | |||
XM_017023871.1 | XP_016879360.1 | 8 | 1090 | |||
XM_017023872.1 | XP_016879361.1 | 8 | 910 | |||
XM_017023873.1 | XP_016879362.1 | 8 | 1077 | |||
XM_017023874.1 | XP_016879363.1 | 8 | 910 | |||
XM_017023875.1 | XP_016879364.1 | 6 | 617 | |||
XM_017023876.1 | XP_016879365.1 | 8 | 955 | |||
XM_017023877.1 | XP_016879366.1 | 8 | 910 | |||
GenBank mRNA | AK022425.1 | - | 8 | 910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
