siRNA Details
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032178.2 | NP_115554.2 | 5 | 1535 | ||
GenBank mRNA | AK023353.1 | - | 5 | 1515 | ||
AK091674.1 | - | 4 | 1637 | |||
AK225712.1 | - | 5 | 1516 | |||
BC013778.1 | - | 5 | 1501 | 4345035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
