siRNA Details
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308068.1 | NP_001294997.1 | 10 | 2596 | ||
NM_032296.2 | NP_115672.2 | 10 | 2593 | |||
XM_006720962.3 | XP_006721025.1 | 12 | 2845 | |||
XM_011522701.2 | XP_011521003.1 | 11 | 3234 | |||
XM_011522702.2 | XP_011521004.1 | 11 | 3708 | |||
XM_011522703.2 | XP_011521005.1 | 12 | 3329 | |||
XM_011522704.2 | XP_011521006.1 | 11 | 3232 | |||
XM_011522705.2 | XP_011521007.1 | 12 | 2990 | |||
XM_017023772.1 | XP_016879261.1 | 13 | 3426 | |||
GenBank mRNA | AB046772.1 | - | 10 | 2573 | ||
AK026885.1 | - | 5 | 762 | |||
AK055777.1 | - | 4 | 1358 | |||
AK057022.1 | - | 5 | 2073 | |||
AK092434.1 | - | 6 | 1413 | |||
AK127235.1 | - | 5 | 2717 | |||
AL136585.1 | - | 8 | 1565 | |||
BC018712.1 | - | 6 | 1341 | 4875281 | ||
BC028572.1 | - | 10 | 2597 | 4839062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
