siRNA Details
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308068.1 | NP_001294997.1 | 10 | 2779 | ||
NM_032296.2 | NP_115672.2 | 10 | 2776 | |||
XM_006720962.3 | XP_006721025.1 | 12 | 3028 | |||
XM_011522701.2 | XP_011521003.1 | 11 | 3417 | |||
XM_011522702.2 | XP_011521004.1 | 11 | 3891 | |||
XM_011522703.2 | XP_011521005.1 | 12 | 3512 | |||
XM_011522704.2 | XP_011521006.1 | 11 | 3415 | |||
XM_011522705.2 | XP_011521007.1 | 12 | 3173 | |||
XM_017023772.1 | XP_016879261.1 | 13 | 3609 | |||
GenBank mRNA | AB046772.1 | - | 10 | 2756 | ||
AK026885.1 | - | 5 | 945 | |||
AK055777.1 | - | 4 | 1541 | |||
AK057022.1 | - | 5 | 2256 | |||
AK092434.1 | - | 6 | 1596 | |||
AK127235.1 | - | 5 | 2900 | |||
AL136585.1 | - | 8 | 1748 | |||
BC018712.1 | - | 6 | 1524 | 4875281 | ||
BC028572.1 | - | 10 | 2780 | 4839062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
