siRNA Details
Gene Symbol: LRRC37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321350.1 | NP_001308279.1 | 12 | 2528 | ||
NM_052888.2 | NP_443120.2 | 9 | 2334 | |||
GenBank mRNA | AJ314647.1 | - | 9 | 2320 | ||
AK299772.1 | - | 12 | 2382 | |||
AK302740.1 | - | 9 | 2465 | |||
BC117373.1 | - | 7 | 2220 | 40125924 | ||
BC143578.1 | - | 8 | 2254 | |||
BC143581.1 | - | 7 | 2220 | 9052092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
