siRNA Details
Gene Symbol: SLC38A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138570.3 | NP_612637.1 | 14 | 2666 | ||
GenBank mRNA | AK093037.1 | - | 8 | 1625 | ||
AK098424.1 | - | 1 | 1194 | |||
BC014642.1 | - | 14 | 2593 | 3028844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
