siRNA Details
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281786.1 | NP_001268715.1 | 2, 3 | 315 | ||
NM_004169.4 | NP_004160.3 | 2, 3 | 315 | |||
NM_148918.2 | NP_683718.1 | 2, 3 | 315 | |||
XM_005256767.3 | XP_005256824.1 | 3, 4 | 386 | |||
XM_011523992.2 | XP_011522294.1 | 2, 3 | 283 | |||
XM_017024957.1 | XP_016880446.1 | 2, 3 | 140 | |||
XM_017024958.1 | XP_016880447.1 | 3, 4 | 386 | |||
XM_017024959.1 | XP_016880448.1 | 1, 2 | 123 | |||
XM_017024960.1 | XP_016880449.1 | 1, 2 | 123 | |||
GenBank mRNA | AK223552.1 | - | 2, 3 | 246 | ||
AK298415.1 | - | 2, 3 | 247 | |||
AK302834.1 | - | 2, 3 | 253 | |||
BC007979.2 | - | 2, 3 | 161 | 3687873 | ||
BC022874.1 | - | 2, 3 | 165 | 4108106 | ||
BC038598.1 | - | 2, 3 | 172 | 4523709 | ||
DB451683.1 | - | 2, 3 | 308 | |||
L11931.1 | - | 1, 2 | 128 | |||
L23928.1 | - | 1, 2 | 116 | |||
Y14485.1 | - | 2, 3 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
