siRNA Details
Gene Symbol: TMEM106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291586.1 | NP_001278515.1 | 5 | 720 | ||
NM_001291587.1 | NP_001278516.1 | 6 | 554 | |||
NM_001291588.1 | NP_001278517.1 | 4 | 530 | |||
NM_145041.3 | NP_659478.1 | 5 | 698 | |||
XM_006721658.2 | XP_006721721.1 | 4 | 525 | |||
XM_006721659.3 | XP_006721722.1 | 6 | 551 | |||
XM_011524260.1 | XP_011522562.1 | 5 | 652 | |||
XM_017024113.1 | XP_016879602.1 | 5 | 652 | |||
XM_017024114.1 | XP_016879603.1 | 5 | 810 | |||
GenBank mRNA | AK056132.1 | - | 5 | 650 | ||
AK290387.1 | - | 5 | 672 | |||
AK299974.1 | - | 6 | 506 | |||
AK301571.1 | - | 4 | 482 | |||
BC012139.2 | - | 4 | 602 | 4562851 | ||
BC146974.1 | - | 4 | 514 | 9056902 | ||
BC146976.1 | - | 4 | 514 | 9056904 | ||
BC147023.1 | - | 4 | 514 | 9056951 | ||
BC157892.1 | - | 4 | 514 | 9057097 | ||
DC419931.1 | - | 4 | 530 |
