siRNA Details
Gene Symbol: WARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004184.3 | NP_004175.2 | 2 | 460 | ||
NM_173701.1 | NP_776049.1 | 2 | 236 | |||
XM_005268044.3 | XP_005268101.1 | 2 | 188 | |||
XM_006720249.3 | XP_006720312.1 | 3 | 422 | |||
XM_011537133.2 | XP_011535435.1 | 4 | 434 | |||
XM_011537135.2 | XP_011535437.1 | 3 | 318 | |||
XM_017021627.1 | XP_016877116.1 | 3 | 326 | |||
XM_017021628.1 | XP_016877117.1 | 3 | 457 | |||
XM_017021629.1 | XP_016877118.1 | 4 | 432 | |||
GenBank mRNA | AK056100.1 | - | 2 | 698 | ||
AK300255.1 | - | 2 | 343 | |||
BC017489.1 | - | 2 | 336 | 3542671 | ||
BC095453.1 | - | 3 | 347 | 30418785 | ||
BM834473.1 | - | 3 | 572 | |||
BX248006.1 | - | 2 | 180 | |||
M61715.1 | - | 2 | 266 | |||
M77804.1 | - | 1 | 121 | |||
X59892.1 | - | 2 | 190 | |||
X62570.1 | - | 1 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
