siRNA Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018100.4 | NP_001018110.1 | 13 | 2316 | ||
NM_152451.7 | NP_689664.3 | 12 | 2232 | |||
GenBank mRNA | AK055535.1 | - | 13 | 2279 | ||
AL833028.1 | - | 8 | 1559 | |||
AL833142.1 | - | 11 | 2013 |
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX095570.1 | - | 1 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
