siRNA Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046357.1 | 2 | 455 | |||
GenBank mRNA | AK055997.1 | - | 3 | 589 | ||
AK074561.1 | - | 2 | 481 | |||
AK091537.1 | - | 1 | 2001 | |||
DA077941.1 | - | 2 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
