siRNA Details
Gene Symbol: SPERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286341.1 | NP_001273270.1 | 2 | 1490 | ||
NM_001286342.1 | NP_001273271.1 | 3 | 1456 | |||
NM_152719.2 | NP_689932.1 | 3 | 1571 | |||
XM_011534971.2 | XP_011533273.1 | 4 | 2926 | |||
GenBank mRNA | AK093129.1 | - | 3 | 1564 | ||
AK292350.1 | - | 2 | 1792 | |||
BC027609.2 | - | 2 | 1390 | 4829821 |
