siRNA Details
Gene Symbol: MEIOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163560.2 | NP_001157032.1 | 12 | 1266 | ||
NM_152764.2 | NP_689977.2 | 12 | 1189 | |||
GenBank mRNA | BC029829.1 | - | 8 | 791 | 5171851 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
