siRNA Details
Gene Symbol: SPECC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033554.2 | NP_001028726.1 | 6 | 2536 | ||
NM_001243438.1 | NP_001230367.1 | 6 | 2545 | |||
NM_152904.4 | NP_690868.3 | 8 | 2627 | |||
XM_011524075.1 | XP_011522377.1 | 7 | 2588 | |||
XM_017025322.1 | XP_016880811.1 | 7 | 2603 | |||
XM_017025323.1 | XP_016880812.1 | 7 | 2612 | |||
XM_017025325.1 | XP_016880814.1 | 8 | 2721 | |||
GenBank mRNA | AB041533.1 | - | 8 | 2627 | ||
AY816326.1 | - | 6 | 2441 | |||
AY816328.1 | - | 8 | 2627 | |||
BC033618.1 | - | 5 | 1651 | |||
BC050058.1 | - | 6 | 2381 | 5277450 | ||
CN361555.1 | - | 1 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
