siRNA Details
Gene Symbol: C15orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301106.1 | NP_001288035.1 | 3 | 1088 | ||
NM_153040.2 | NP_694585.1 | 3 | 1050 | |||
GenBank mRNA | AK057393.1 | - | 3 | 1051 | ||
BC109252.2 | - | 3 | 844 | 40023840 | ||
BC109253.1 | - | 3 | 804 | 40023844 |
