siRNA Details
Gene Symbol: CA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289470.1 | - | 3 | 575 | ||
AK293999.1 | - | 2 | 194 | |||
AY075022.1 | - | 3 | 580 | |||
BC069744.1 | - | 3 | 572 | 7262540 | ||
L04656.1 | - | 3 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
