siRNA Details
Gene Symbol: CCDC78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022931.1 | XP_016878420.1 | 1 | 893 | ||
GenBank mRNA | AK127495.1 | - | 4 | 2446 | ||
AK128538.1 | - | 4 | 2851 | |||
AK128575.1 | - | 4 | 2777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
